Learn More
Enzo Life SciencesSupplier Diversity Partner HSP 90 monoclonal antibody (2D12) (50 µl)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADISPA845D
Clone: 2D12. Host: Rat. Isotype: IgM. Immunogen: Native mouse Hsp90. UniProt: P07901 (HSP90alpha), P11499 (HSP90beta). Source: Purified from mouse ascites. Species Reactivity: Human, Mouse, Rat, Water mold, Sheep, Rabbit, Monkey, Hamster, Guinea pig, Fish, Dog, Chicken, Bovine, Beluga. Applications: IP, WB. Recommended dilutions: Western Blot (1:500, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Ammonium sulfate precipitation purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-638