Learn More
HSP90 beta Mouse anti-Human, DyLight 755, Clone: 8D3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11096428IR
Catalog No. NBT96428IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
HSP90 beta
8D3
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
HSP90AB1
Ah receptor (Aryl hydrocarbon receptor)
PEG purified
Cancer, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific
Primary
3326
Human
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, heat shock protein 90kDa alpha (cytosolic), class B member 1, heat shock protein beta, heat shock protein HSP 90-beta, HSP 84, HSP84, HSP90-BETA, HSP90BHSP 90, HSPC2D6S182, HSPCBFLJ26984
Mouse
IgM
0.2 ml
Store at 4C in the dark.
Monoclonal
Detects 90kDa. Co-immunoprecipitates HSP90 complexes, including HSP70, Hop, Ah receptors, glucocorticoid receptors, heme-regulated eukaryotic initiation factor 2a (eIF-2a) kinase (HRI).