Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90 beta Mouse anti-Human, Mouse, Alexa Fluor 647, Clone: Hyb-K3701, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11096871AF647

Catalog No. NBT96871647

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

HSP90 beta
Hyb-K3701
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
HSP90AB1
Recombinant human HSP90beta; Specificity mapped to amino acids 185-335
Protein G purified
Cancer, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific
Primary
3326
Human, Mouse
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, heat shock protein 90kDa alpha (cytosolic), class B member 1, heat shock protein beta, heat shock protein HSP 90-beta, HSP 84, HSP84, HSP90-BETA, HSP90BHSP 90, HSPC2D6S182, HSPCBFLJ26984
Mouse
IgM
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects 90kD proteins corresponding to the molecular mass of hsp90beta.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit