Learn More
HSP90 Mouse anti-Achlya, DyLight 350, Clone: AC-16, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11096430UV
Catalog No. NBT96430UV
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
HSP90
AC-16
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
HSP90AA1
Heat shock protein 90 from the water mold Achyla ambisexualis
Protein G purified
Cancer, Hypoxia, Membrane Trafficking and Chaperones
Primary
3320
Achlya
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Mouse
IgG2b
0.2 ml
Store at 4C in the dark.
Monoclonal
Detects 90kDa. This antibody is reactive with both the constitutive and the inducible form of HSP90. It does not bind to the native form and does not recognize HSP90 from E.coli or yeast.