Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90 Mouse anti-Bovine, Chicken, Human, Mouse, Porcine, Rabbit, Rat, Clone: D7A, Novus Biologicals™

Mouse MonoclonalAntibody

$153.00 - $358.00

Specifications

Antigen HSP90
Clone D7A
Dilution Western Blot 1:500, Immunohistochemistry, Immunoprecipitation 5ug, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
NB59683100U View Documents Novus BiologicalsSupplier Diversity Partner
NBP259683100UG
100 ug Unconjugated Each for $358.00
Add to cart
 
NB5968325UG View Documents Novus BiologicalsSupplier Diversity Partner
NBP25968325UG
25 ug Unconjugated Each for $153.00
Add to cart
 
Description

Description

HSP90 Monoclonal antibody specifically detects HSP90 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

HSP90
Western Blot 1:500, Immunohistochemistry, Immunoprecipitation 5ug, Immunohistochemistry-Paraffin
Purified
Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
Full length protein HSP90 purified from chicken brain
Monoclonal
Protein G purified
Can isolate complexes of HSP90, Src kinase and cec37.
D7A
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Mouse
RUO
3320
IgG
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit