Learn More
HSP90 Mouse anti-Chicken, Alexa Fluor 405, Clone: D7A, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11096872AF405
Catalog No. NBT96872405
Add to cart
Description
Description
HSP90 Monoclonal antibody specifically detects HSP90 in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
HSP90
D7A
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
P11501
FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Recognizes 90kDa. Can isolate complexes of HSP90, Src kinase and cec37.
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
HSP90AA1
Full length protein HSP90 purified from chicken brain
Protein A or G purified
Cancer, Hypoxia, Membrane Trafficking and Chaperones
Primary
3320
Chicken