Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hsp90A/B Mouse anti-Human, Mouse, Rabbit, Clone: H90-10, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Hsp90A/B |
---|---|
Clone | H90-10 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP197529
|
Novus Biologicals
NBP197529 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Hsp90A/B Monoclonal antibody specifically detects Hsp90A/B in Human, Mouse, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
Hsp90A/B | |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human, Mouse, Rabbit | |
3320 | |
Purified human HSP90beta (heat shock protein 90beta). | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
H90-10 | |
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
HSP90AA1 | |
IgG2a | |
Protein G purified | |
Recognizes human, mouse and rabbit HSP90 alpha and HSP90 beta. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title