Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp90A/B Mouse anti-Human, Mouse, Rat, Yeast, DyLight 755, Clone: Hyb-K41220A, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11096870IR

Catalog No. NBT96870IR


Add to cart

Description

Description

Hsp90A/B Monoclonal antibody specifically detects Hsp90A/B in Human, Mouse, Rat, Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Hsp90A/B
Hyb-K41220A
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
HSP90AA1
Recombinant human HSP90alpha; Specificity mapped to amino acids 291-304
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 90kD proteins corresponding to the molecular mass of hsp90 alpha or beta.
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Mouse
IgG2a
0.1 ml
Primary
3320
Human, Mouse, Rat, Yeast
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit