Learn More
Aviva Systems Biology Corp HSP90AB1 Antibody - N-terminal region (AVARP09016_T100) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP09016_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HSP90AB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HSP90AB1. Alternate names: HSPC2, HSPCB, D6S182, HSP90B, HSP90-BETAApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse
Catalog No. 50-014-62569