Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA12B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548597

Catalog No. PA548597

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

The protein encoded by this gene contains an atypical heat shock protein 70 (Hsp70) ATPase domain and is therefore a distant member of the mammalian Hsp70 family. This gene may be involved in susceptibility to atherosclerosis. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Dec 2015]
Specifications

Specifications

HSPA12B
Polyclonal
PBS with 0.09% sodium azide
Q96MM6
Heat shock 70 kDa protein 12B, HSPA12B, C20orf60
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
116835
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HSPA12B
Liquid
HSPA12B
KLH conjugated synthetic peptide between 108-136 amino acids from the N-terminal region of human HSPA12B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit