Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA4 (Human) Recombinant Protein (P01), Abnova

Human HSPA4 full-length ORF with GST-tag at N-terminal

$403.50 - $627.50

Specifications

Cross Reactivity Human
Format Solution
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 42.02
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-020-339
View Documents
Abnova Corporation
H00003308P0110
10 ug Each for $403.50
Add to cart
 
89-020-356
View Documents
Abnova Corporation
H00003308P0125
25 ug Each for $627.50
Add to cart
 
Description

Description

Heat shock 70kDa protein 4

  • Theoretical MW: 42.02kDa
  • Preparation Method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Best use within three months from the date of receipt of this protein

Enzyme Linked Immunosorbent Assay, Western Blotting, Antibody Production, Protein Array

Specifications

Specifications

Human
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
42.02
8
Glutathione Sepharose 4 Fast Flow
Yes
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Solution
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
HSPA4 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit