Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspA4 Mouse anti-Amphibian, Bacteria, Brine Shrimp, Chicken, Drosophila, Fish, Human, Mouse, Rat, Yeast, Clone: 3A3, Novus Biologicals™

Mouse MonoclonalAntibody

$204.00 - $308.00

Specifications

Antigen HspA4
Clone 3A3
Dilution Western Blot 1:5000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 2ug
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
NB59686100U View Documents Novus BiologicalsSupplier Diversity Partner
NBP259686100UG
100 ug Unconjugated Each for $308.00
Add to cart
 
NB5968625UG View Documents Novus BiologicalsSupplier Diversity Partner
NBP25968625UG
25 ug Unconjugated Each for $204.00
Add to cart
 
Description

Description

HspA4 Monoclonal antibody specifically detects HspA4 in Human, Mouse, Rat, Amphibian, Bacteria, Brine Shrimp, Chicken, Drosophila, Fish, Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

HspA4
Western Blot 1:5000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 2ug
Purified
Amphibian, Bacteria, Brine Shrimp, Chicken, Drosophila, Fish, Human, Mouse, Rat, Yeast
PBS pH7.2, 50% glycerol with 0.09% Sodium Azide
Human recombinant HSP70 overexpressed in E.coli
Monoclonal
Protein G purified
May detect HSP70, HSC70, p75 and HSP72.
3A3
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Mouse
RUO
3308
IgG1
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit