Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSPA4L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558671
Description
Immunogen sequence: KCHAEHTPEE EIDHTGAKTK SAVSDKQDRL NQTLKKGKVK SIDLPIQSSL CRQLGQDLLN SY Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
HSPA4L possesses chaperone activity in vitro where it inhibits aggregation of citrate synthase.
Specifications
HSPA4L | |
Polyclonal | |
Unconjugated | |
HSPA4L | |
94kDa; AI461691; APG1; APG-1; heat shock 70 kDa protein 4L; heat shock 70 kDa protein 4-like protein; heat shock 70 kDa protein 4L-like; Heat shock 70 kDa protein 4L-like protein; heat shock 70kDa protein 4 like; heat shock 70kDa protein 4-like; heat shock 70-related protein APG-1; Heat shock protein; heat shock protein (hsp110 family); heat shock protein 4 like; heat shock protein 4-like; heat shock protein family A (Hsp70) member 4 like; HSP; Hsp4l; HSPA4L; HSPH3; LOW QUALITY PROTEIN: heat shock 70 kDa protein 4L; osmotic stress protein 94; OSP94; testis tissue sperm-binding protein Li 64n | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22824 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95757 | |
HSPA4L | |
Recombinant protein corresponding to Human HSPA4L. Recombinant protein control fragment (Product #RP-104170). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction