Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA4L Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549636

Catalog No. PA549636

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HSPA4L possesses chaperone activity in vitro where it inhibits aggregation of citrate synthase.
Specifications

Specifications

HSPA4L
Polyclonal
PBS with 0.09% sodium azide
O95757
Heat shock 70 kDa protein 4L, Heat shock 70-related protein APG-1, Osmotic stress protein 94, HSPA4L, APG1, OSP94
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22824
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HSPA4L
Liquid
HSPA4L
KLH conjugated synthetic peptide between 776-803 amino acids from the C-terminal region of human HSPA4L.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit