Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA5 Mouse anti-Human, Rat, Clone:9E4-2A7-H6, Abnova™

Mouse monoclonal antibody raised against recombinant human HSPA5.

Manufacturer:  Abnova Corporation MAB17662

Catalog No. 89-333-884


Add to cart

Description

Description

When Chinese hamster K12 cells are starved of glucose, the synthesis of several proteins, called glucose-regulated proteins (GRPs), is markedly increased. Hendershot et al. (1994) [PubMed 8020977] pointed out that one of these, GRP78 (HSPA5), also referred to as  ft.immunoglobulin heavy chain-binding protein ft. (BiP), is a member of the heat-shock protein-70 (HSP70) family and is involved in the folding and assembly of proteins in the endoplasmic reticulum (ER). Because so many ER proteins interact transiently with GRP78, it may play a key role in monitoring protein transport through the cell.[supplied by OMIM]
Specifications

Specifications

HSPA5
9E4-2A7-H6
heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa)
In PBS, pH 7.4 (50% glycerol, 0.02% sodium azide)
Liquid
Mouse
IgG1
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This antibody detects endogenous levels of HSPA5 and does not cross-react with related proteins.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Immunocytochemistry (1:50)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
HSPA5
BIP, FLJ26106, GRP78, MIF2
Recombinant protein corresponding to a fragment at C-terminus of human HSPA5 from E. coli.
100μg
Antibody
3309
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit