Learn More
Aviva Systems Biology Corp HSPA6 Antibody - middle region (ARP54649_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54649_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPA6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSPA6. Alternate names: -ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-61274