Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HspA6 Mouse anti-Human, PerCP, Clone: OTI1H3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270959PCP
Description
HspA6 Monoclonal specifically detects HspA6 in Human samples. It is validated for Flow Cytometry.Specifications
HspA6 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
HSPA6 | |
Full length recombinant protein of human HspA6 (NP_002146) produced in HEK293T cells. | |
0.1 mL | |
3310.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI1H3 | |
Flow Cytometry | |
heat shock 70 kDa protein 6, Heat shock 70 kDa protein B′, heat shock 70kD protein 6 (HSP70B′), heat shock 70kDa protein 6 (HSP70B′), HSP70B′ | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title