Learn More
Invitrogen™ HSPB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579411
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human U20S whole cell, human PC-3 whole cell, rat H9C2 whole cell, rat heart tissue, mouse C2C12 whole cell, mouse heart tissue IHC-F: Rat Cardiac Muscle Tissue. ICC/IF: HELA Cell.
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The protein is expressed preferentially in the heart and skeletal muscle. This protein regulates Myotonic Dystrophy Protein Kinase, which plays an important role in maintenance of muscle structure and function.
Specifications
HSPB2 | |
Polyclonal | |
Unconjugated | |
HSPB2 | |
27kDa; 2810021G24Rik; DMPK-binding protein; heat shock 27kD protein 2; heat shock 27kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein B2; heat shock protein beta 2; heat shock protein beta-2; heat shock protein family B (small) member 2; heat-shock protein beta-2; Hs.78846; HSP; HSP27; Hspb2; LOH11CR1K; MGC133245; MKBP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
161476, 3316, 69253 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
O35878, Q16082, Q99PR8 | |
HSPB2 | |
A synthetic peptide corresponding to a sequence in the middle region of human HSPB2 (102-118aa RHPQRLDRHGFVSREFC). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.