Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspB7 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen HspB7
Immunogen Synthetic peptides corresponding to Hspb7 (heat shock protein family, member 7 (cardiovascular)) The peptide sequence was selected from the C terminal of Hspb7. Peptide sequence TFAHKCQLPEDVDPTSVTSALREDGSLTIRARRNPHTEHVQQTFRTEIKI.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1690720 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16907120UL
20ul Each for $149.00
Add to cart
 
NBP169071 View Documents Novus BiologicalsSupplier Diversity Partner
NBP169071
100 ul Each for $329.00
Add to cart
 
Description

Description

HspB7 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

HspB7
Rabbit
Polyclonal
Unconjugated
27129
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to Hspb7 (heat shock protein family, member 7 (cardiovascular)) The peptide sequence was selected from the C terminal of Hspb7. Peptide sequence TFAHKCQLPEDVDPTSVTSALREDGSLTIRARRNPHTEHVQQTFRTEIKI.
Primary
IgG
Cardiovascular heat shock protein, CVHSP, DKFZp779D0968, FLJ32733, heat shock 27kDa protein family, member 7 (cardiovascular), heat shock protein beta-7, HspB7, member 7 (cardiovascular)
PBS & 2% Sucrose. with No Preservative
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit