Learn More
HSPB8/HSP22 Mouse anti-Human, Mouse, Rat, DyLight 550, Clone: 3C12-H11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP148303R
Catalog No. NBP148303R
Add to cart
Description
Description
HSPB8/HSP22 Monoclonal antibody specifically detects HSPB8/HSP22 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
HSPB8/HSP22
3C12-H11
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
HSPB8
His-tagged human recombinant HSP22
Protein G purified
Protein Kinase
Primary
26353
Human, Mouse, Rat
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Alpha-crystallin C chain, CRYAC, E2IG1E2-induced gene 1 protein, H11DHMN2, heat shock 22kDa protein 8, heat shock 27kDa protein 8, heat shock protein beta-8, HMN2, HMN2A, HSP22CMT2L, HspB8, Protein kinase H11, Small stress protein-like protein HSP22
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 22kDa. Does not cross react with alpha crystallin.