Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPB8/HSP22 Mouse anti-Human, Rat, PE/Atto594, Clone: OTI1E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271828PEATT594

Catalog No. N271828P594


Add to cart

Description

Description

HSPB8/HSP22 Monoclonal antibody specifically detects HSPB8/HSP22 in Human, Rat samples. It is validated for Flow Cytometry.
Specifications

Specifications

HSPB8/HSP22
OTI1E3
Flow Cytometry
Affinity Purified, Conjugated
HSPB8
Full length recombinant protein of human HSPB8/HSP22 (NP_055180) produced in HEK293T cells.
Immunogen affinity purified
Protein Kinase
Primary
26353.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
Alpha-crystallin C chain, CRYAC, E2IG1E2-induced gene 1 protein, H11DHMN2, heat shock 22kDa protein 8, heat shock 27kDa protein 8, heat shock protein beta-8, HMN2, HMN2A, HSP22CMT2L, HspB8, Protein kinase H11, Small stress protein-like protein HSP22
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit