Learn More
HSPB8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152871AP
Catalog No. 50-173-0123
Add to cart
Description
Description
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The expression of this gene in induced by estrogen in estrogen receptor-positive breast cancer cells, and this protein also functions as a chaperone in association with Bag3, a stimulator of macroautophagy. Thus, this gene appears to be involved in regulation of cell proliferation, apoptosis, and carcinogenesis, and mutations in this gene have been associated with different neuromuscular diseases, including Charcot-Marie-Tooth disease.
Specifications
Specifications
HSPB8
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9EPX0, Q9JK92, Q9UJY1
Alpha crystallin C chain, CMT2L, CRYAC, DHMN2, E2 induced gene 1 protein, E2IG1, H11, heat shock 22kDa protein 8, Heat shock protein beta 8, HMN2, HMN2A, HSP22, HSPB8, Protein kinase H11
Rabbit
IgG
150 μL
-20°C
Primary
113906, 26353, 80888
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HSPB8
Liquid
Hspb8
HSPB8 Fusion Protein Ag7456
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat