Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspBAP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18002720UL

 View more versions of this product

Catalog No. NBP18002720


Add to cart

Description

Description

HspBAP1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

HspBAP1
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
HSPBAP1
Synthetic peptide directed towards the middle region of human HSPBAP1. Peptide sequence GCNLVFQVQGRKRWHLFPPEDTPFLYPTRIPYEESSVFSKINVVNPDLKR.
55 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:1000
NP_078886
FLJ22623, heat shock 27 kDa associated protein, HSPB (heat shock 27kD) associated protein 1, HSPB (heat shock 27kDa) associated protein 1, HSPB1-associated protein 1,27 kDa heat shock protein-associated protein 1, PASS1FLJ39386, Protein associated with small stress protein 1, protein associating with small stress protein PASS1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
79663
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit