Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HspBP1 Mouse anti-Human, Monkey, Allophycocyanin/Cy7, Clone: OTI1D5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271761APCCY7

Catalog No. N271761APC7


Add to cart

Description

Description

HspBP1 Monoclonal antibody specifically detects HspBP1 in Human, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

HspBP1
OTI1D5
Flow Cytometry
Affinity Purified, Conjugated
HSPBP1
Full length recombinant protein of human HspBP1 (NP_036399) produced in HEK293T cells.
Immunogen affinity purified
Signal Transduction
Primary
23640.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
FES1, Heat shock protein-binding protein 1, Hsp70 binding protein 1, hsp70 interacting protein, hsp70-binding protein 1, Hsp70-binding protein 2, Hsp70-interacting protein 1, Hsp70-interacting protein 2, HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1, HSPBP, HspBP1, hspBP2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit