Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPBP1 Monoclonal Antibody (OTI1F3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA525667
Description
HSPBP1 Monoclonal antibody specifically detects HSPBP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
HSPBP1 | |
Monoclonal | |
0.58 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NZL4 | |
HSPBP1 | |
Full length human recombinant protein of HSPBP1 produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1F3 | |
Unconjugated | |
HSPBP1 | |
1500019G21Rik; FES1; Heat shock protein; heat shock protein-binding protein 1; HSP; hsp70 interacting protein; hsp70-binding protein 1; Hsp70-binding protein 2; hsp70-interacting protein; hsp70-interacting protein 1; Hsp70-interacting protein 2; HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1; HSPA (Hsp70) binding protein 1; HSPA binding protein, cytoplasmic cochaperone 1; Hspbp; HSPBP1; HspBP2; LOW QUALITY PROTEIN: hsp70-binding protein 1; MGC128574 protein; PP1845; SPA ) binding protein, cytoplasmic cochaperone 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23640 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction