Learn More
HSPC111 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00051491T01
Catalog No. 89-002-234
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
BC040106
6.8
Transient overexpression cell Lysate was tested with Anti-HSPC111 antibody (H00051491-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
HSPC111 293T Cell Transient Overexpression Lysate(Denatured)
19.69
pCMV-HSPC111 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.