Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BABAM1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549611

Catalog No. PA549611

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

Component of the BRCA1-A complex, a complex that specifically recognizes  ft.Lys-63 ft.-linked ubiquitinated histones H2A and H2AX at DNA lesions sites, leading to target the BRCA1-BARD1 heterodimer to sites of DNA damage at double-strand breaks (DSBs). The BRCA1-A complex also possesses deubiquitinase activity that specifically removes  ft.Lys-63 ft.-linked ubiquitin on histones H2A and H2AX. In the BRCA1-A complex, it is required for the complex integrity and its localization at DSBs. Probably also plays a role as a component of the BRISC complex, a multiprotein complex that specifically cleaves  ft.Lys-63 ft.-linked ubiquitin. In these 2 complexes, it is probably required to maintain the stability of BRE/BRCC45 and help the  ft.Lys-63 ft.-linked deubiquitinase activity mediated by BRCC3/BRCC36 component.
Specifications

Specifications

BABAM1
Polyclonal
PBS with 0.09% sodium azide
Q9NWV8
BRISC and BRCA1-A complex member 1, Mediator of RAP80 interactions and targeting subunit of 40 kDa, New component of the BRCA1-A complex, BABAM1, C19orf62, MERIT40, NBA1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29086
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
BABAM1
Liquid
BABAM1
KLH conjugated synthetic peptide between 116-143 amino acids from the Central region of human HSPC142.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit