Learn More
Aviva Systems Biology Corp HSPE1 Antibody - middle region (ARP54651_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54651_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSPE1. Alternate names: CPN10, GROES, HSP10, EPFApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61275