Learn More
HSPG2 Rat anti-Human, Clone: A7L6, Abnova™
Rat monoclonal antibody raised against native human HSPG2.
Manufacturer:  Abnova Corporation MAB12489
Catalog No. 89-934-319
Add to cart
Description
Description
Heparan sulfate proteoglycan is a major component of basement membranes, where the molecule may be involved in the stabilization of other molecules as well as being involved with glomerular permeability to macromolecules and cell adhesion. This form of HSPG, known as HSPG2 or perlecan, is encoded by a gene that maps to chromosome 1. The gene for the form of HSPG associated with the cell surface of fibroblasts has been mapped to human chromosome 8 (MIM 142460).[supplied by OMIM]
Specifications
Specifications
HSPG2
A7L6
Rat monoclonal antibody raised against native human HSPG2.
In PBS (0.09% sodium azide)
PLC/PRCAN/SJA/SJS/SJS1
Rat
IgG2a
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Unconjugated
Immunocytochemistry Immunohistochemistry (Frozen sections) (1:50) Immunoprecipitation (1:100-1:1000) Western Blot The optimal working dilution should be determined by the end user.
Liquid
HSPG2
High molecular mass material derived from the Engelbreth-Holm-Swarm (EHS) tumor matrix containing laminin, entactin and HSPG.
Protein G purification
RUO
Primary
3339