Learn More
Hspg2 Rat anti-Human, Mouse, Clone: A7L6, Abnova™
Rat monoclonal antibody raised against native mouse Hspg2.
Manufacturer:  Abnova Corporation MAB13313
Catalog No. 89-934-317
Add to cart
Description
Description
Specifications
Specifications
Hspg2
A7L6
Rat monoclonal antibody raised against native mouse Hspg2.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
AI852380/Pcn/Plc/per
Rat
IgG2a, kappa
100 ug
Store at 4°C.
Monoclonal
Human, Murine
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Hspg2
Murine EHS laminin preparation.
Protein A/G purification
RUO
Primary
15530