Learn More
Drg International IncSupplier Diversity Partner Hsv-1+2 Igg Elisa Ruo DFS Item
Manufacturer:  Drg International IncSupplier Diversity Partner EIA3489R
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to herpes simplex virus type 1+ 2 in serum and plasma. This product is for Research Use Only. The DRG Herpes Simplex Virus Type 1+2 IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with recombinant gG1 Protein of HSV-1 and native gG2 protein of HSV-2. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Herpes Simplex Virus Type 1+2-specific antibodies of positive specimens and controls are bound to the immobilized antigens.
Catalog No. 50-194-7577