Learn More
Drg International IncSupplier Diversity Partner Hsv-2 Igm Elisa Ruo DFS Item
Manufacturer:  Drg International IncSupplier Diversity Partner EIA3488R
An enzyme immunoassay for the qualitative and semiquantitative determination of IgM-class antibodies to HSV-2 in serum. This product is for Research Use Only. The DRG Herpes Simplex Virus Type 2 IgM ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Patient samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, containing hyper-immune anti-human IgG-class antibody to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors. This pretreatment avoids false negative or false positive results.
Catalog No. 50-194-7576