Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ HSV Tag Monoclonal Antibody,
Mouse Monoclonal Antibody
$270.00 - $1033.00
Specifications
Antigen | HSV Tag |
---|---|
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Host Species | Mouse |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
69-171-3MI
|
MilliporeSigma™
691713 |
Unconjugated | 40 μg |
Each for $270.00
|
|
||||
69-171-4MI
|
MilliporeSigma™
691714 |
Unconjugated | 200 μg |
Each for $1,033.00
|
|
||||
Description
The HSV Tag™ Antibody is a mouse monoclonal antibody (IgG1) that has high specificity and affinity for an 11-amino acid peptide derived from Herpes Simplex Virus (HSV) glycoprotein D. The peptide is encoded by the HSV Tag sequence in appropriate expression vectors and by the Positive Control 36mer in the Novagen NovaTope™ System. The purified antibody produces a strong signal at a 1:10,000 dilution on Western blots (Sakai 1996), dot blots, and colony lifts when used with the Novagen Anti-Mouse IgG AP Conjugate.Specifications
HSV Tag | |
Unconjugated | |
Mouse | |
Stabilized solution of 1mg/mL of pure antibody in 50% glycerol, includes positive control extract containing 31.1kDa Protein | |
Primary | |
HSV tag peptide GlnProGluLeuAlaProGluAspProGluAsp , C-terminal or internal |
Monoclonal | |
Purified | |
RUO | |
IgG1 | |
-20°C |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title