Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSV Tag Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA126555

Catalog No. PIPA126555


Add to Cart

Description

Description

PA1-26555 detects HSV tag.

HSV Tag Polyclonal antibody specifically detects HSV Tag in Tag samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSV Tag
Polyclonal
Unconjugated
Herpes Simplex 2; Herpes Simplex II; HHV Epitope tag; HHV-2; HSV epitope tag; HSV Type 2; HSV Type II; HSV2; HSV-2
Synthetic peptide, conjugated to KLH, corresponding to residues Q P E L A P E D P E D C of HSV.
100 μg
Primary
4° C
Liquid
Tag
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Rabbit
Antigen affinity chromatography
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit