Learn More
Invitrogen™ HSV Type 1 gD Monoclonal Antibody (1-I-9)
Description
MA1-7594 detects HSV from virus samples. MA1-7594 has been successfully used in ELISA, Immunoprecipitation, immunofluorescence, and Western blot procedures. The MA1-7594 immunogen is a HSV (I) viral concentrate.
Specifications
Specifications
| Antigen | HSV Type 1 gD |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1-I-9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.01% sodium azide; pH 7.2 |
| Gene Alias | Herpes Simplex 1; Herpes Simplex I; HHV; HSV; HSV Type 1; HSV1 |
| Host Species | Mouse |
| Immunogen | HSV (I) infected cells. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.