Learn More
Invitrogen™ HSV Type 2 gB Monoclonal Antibody (1-G-10)
Description
MA1-7595 detects HSV from virus samples. MA1-7595 has been successfully used in ELISA, Western blot and Immunofluorescence procedures. The MA1-7595 immunogen is HSV (II) infected cells. Store at 4ºC short term. For extended storage aliquot and store at -80ºC or below. Avoid freeze-thaw cycles.
Specifications
Specifications
| Antigen | HSV Type 2 gB |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1-G-10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.01% sodium azide; pH 7.2 |
| Gene Alias | Herpes Simplex 2; Herpes Simplex II; HHV-2; HSV Type 2; HSV Type II; HSV2; HSV-2 |
| Host Species | Mouse |
| Immunogen | HSV (II) infected cells. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.