Learn More
R&D Systems™ HT F Homogeneous PARP Inhibition Assay Kit
HT Fluorescent Screening Assay for PARP Inhibitors
Manufacturer:  R&D Systems™ 4690096K
Catalog No. 46-900-96K
Add to cart
This item is not returnable. View return policy
Description
Description
Kit Contents:
  • 10X Cycling Enzymes (50% glycerol) (600 µL)
  • 2 µM NAD (3.5 mL)
  • PARP I Enzyme, 200 ng/µL (100 µL)
  • Activated DNA, 200 ng/µL (600 µL)
  • 10X Resazurin (600 µL)
  • 10X Buffer (1M Tris-HCl, pH 8) (3.5 mL)
  • Stop Solution (6 mL)
  • 96-well Black Microplate (1 plate)
Specifications
Specifications
96 Tests
Store the unopened product at -20 to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.