Learn More
Trajan™ HT GC Septa
HT GC septa is a bleed and temperature optimized material
Supplier:  Trajan™ 041881
 View more versions of this product
Catalog No. SG041881
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Bleed and temperature optimized septa
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 400°C (330°C for 17mm size)
Specifications
Specifications
BTO Silicone
9.5 mm
350°C
100/Pk.