Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTL Strefa Acti-Lance Safety Lancets

Available in three color-coded models to meet specific blood sample applications and address the variety of patient skin types

Manufacturer:  HTL STREFA INC 7141

 View more versions of this product

Catalog No. 22-192-005

  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Safety Lancet
0.05 in.
Yes
Lite
28g
Yes
Purple
1.5mm
For blood glucose monitoring purposes and is especially recommended for all patients with delicate skin including children
Silicone
200 Pack
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit