Learn More
BioworldSupplier Diversity Partner HTLV-1 gp46 (Recombinant), 100 μg DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220602261
Recombinant HTLV-I gp46 (162-214/242-257aa) Source: E.coli Description: E.coli derived recombinant. The protein contains the gp46 immunodominant regions (162-214/242-257aa). Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. Protein conc: 1 mg/ml containing 10mM Tris-HCl, pH 4.5, 100mM Sodium Phosphate, 8M urea. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -200C. Stability : Five years frozen. One month in solution at room temperature. Applications: Use as an antigen in ELISA and Western Blots. Excellent reagent for correct detection of HTLV infections, with minimal specificity problems. Usage: BioWORLD\\\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-327