Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HTR1F Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551066

Catalog No. PA551066


Only null left
Add to Cart

Description

Description

HTR1F / 5-HT-1F Polyclonal Antibody detects endogenous levels of HTR1F / 5-HT-1F protein.

5-HT1F is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity. 5-HT1F is a member of the GPCR family. Expression of 5-HT1F has been reported in various brain structures, including cortex, hippocampus, trigeminal ganglia, and cerebral blood vessels. Expression in peripheral tissues is limited to uterus, mesentery, and artery. No expression has been detected in heart, kidney, liver, pancreas, or spleen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HTR1F
Polyclonal
Unconjugated
HTR1F
5-HT-1E-beta; 5ht1f; 5-HT1F; 5-HT-1F; 5HT1F Receptor; 5-HT1f receptor; 5HT6; 5-hydroxytryptamine (serotonin) receptor 1E beta; 5-hydroxytryptamine (serotonin) receptor 1F; 5-hydroxytryptamine (serotonin) receptor 1F, G protein-coupled; 5-hydroxytryptamine receptor 1F; Htr1eb; HTR1EL; HTR1F; MR77; Serotonin receptor 1F; serotonin receptor gene (similar to Mm Htr1eb)
Rabbit
Antigen Affinity Chromatography
RUO
15557, 3355, 60448
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P30939, P30940, Q02284
HTR1F
Synthesized peptide derived from the Internal region of human SR-1F (aa 101-150).
100 μL
Primary
Human, Mouse, Rat, Monkey
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.