Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTRA4, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549263

Catalog No. PA549263

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Serine protease.

Specifications

Specifications

HTRA4
Polyclonal
PBS with 0.09% sodium azide
P83105
Serine protease HTRA4, 3421-, High-temperature requirement factor A4, HTRA4
Rabbit
IgG
400μL
Store at 2° to 8°C for up to 6 months. For long term storage store at -20°C.
Primary
203100
Western Blot
Unconjugated
HTRA4
Liquid
HTRA4
KLH conjugated synthetic peptide between 283-311 amino acids from the Central region of human HTRA4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit