Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD196 Mouse anti-Human, Brilliant Ultraviolet 737, Clone: 11A9, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 612780
Description
Ensure accurate, reproducible results in Flow cytometrySpecifications
CD196 | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
11A9 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human CD196/CCR6 Peptide | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction