Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD298 Mouse anti-Human , PE, Clone: P-3E10, BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566792
Description
CD298 Monoclonal antibody specifically detects CD298 in Human samples. It is validated for Flow CytometrySpecifications
CD298 | |
Monoclonal | |
PE | |
ATP1B3; ATPB-3; Na, K-ATPase beta-3 polypeptide; CD298 | |
Human MOLT-4 Cell Line | |
RUO | |
Human | |
IgG2a κ |
Flow Cytometry | |
P-3E10 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction