Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RA Mouse anti-Human, BUV737, Clone: HI100, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 612847
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Flow Cytometry.Specifications
CD45RA | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
25 Tests | |
Primary | |
Human | |
IgG2b κ |
Flow Cytometry | |
HI100 | |
Mouse | |
RUO | |
CD45RA | |
Refrigerate (2 to 8°C) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction