Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR5 (CD185) Rat anti-Human, BUV563, Clone: RF8B2, BD Biosciences™
Rat Monoclonal Antibody
Supplier: BD Biosciences 741316
Description
CXCR5 (CD185) Monoclonal antibody specifically detects CXCR5 (CD185) in Human samples. It is validated for Flow CytometrySpecifications
CXCR5 (CD185) | |
Monoclonal | |
Brilliant Ultraviolet 563 | |
CD185; Chemokine C-X-C motif receptor 5; BLR1; MDR-15 | |
Human CXCR5 (CD185) | |
RUO | |
Human | |
IgG2b κ |
Flow Cytometry | |
RF8B2 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction