Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-9 Receptor Alpha (CD129) Mouse anti-Human, BV605, Clone: AH9R7, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 745142
Description
IL-9 Receptor Alpha (CD129) Monoclonal antibody specifically detects IL-9 Receptor Alpha (CD129) in Human samples. It is validated for Flow Cytometry.Specifications
IL-9 Receptor Alpha (CD129) | |
AH9R7 | |
Brilliant Violet 605 | |
Mouse | |
Monoclonal | |
50 μg | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
0.2 mg/mL | |
Purified | |
IgG2b, κ | |
Primary | |
RUO | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title