Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAG-3 (CD223) Mouse anti-Human, PE-CF594, Clone: T47-530, BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565719
Description
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometrySpecifications
LAG-3 (CD223) | |
Monoclonal | |
PE-CF594 | |
LAG3; CD223; FDC; Lymphocyte activation gene 3 protein; Protein FDC | |
Human LAG-3 Recombinant Protein | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
T47-530 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD HorizonTM PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction