Learn More
Heidolph™ Huber Thermofluids - SilOil
SDP
Heidolph™ Huber Thermofluids - SilOil is recommended for most temperature control applications
Supplier:  Heidolph™ 015950259
Catalog No. 05-405-333
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
  • Huber thermofluids are recommended for temperature control applications
  • Equipped with the best possible thermodynamic and environmental characteristics
  • Safe reliable operation relies on compliance with safety standards to ensure optimal results
Specifications
Specifications
Heat Transfer Fluid
-40°C to +165/220°C
10 L
Temperature Control Applications
10 mm2/sec. (at 25°C)
Recommendations
Recommendations