Learn More
ACROBiosystems Biotinylated Human HLA-A*03:01&B2M Complex Protein (Peptide free, MALS verified)
Supplier:  ACROBiosystems HLMH82W625UG
Encompass_Preferred
Biotinylated Human HLA-A*03:01&B2M Complex Protein (HLM-H82W6) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*03:01) & Ile 21 - Met 119 (B2M) (Accession # P04439-1 (HLA-A*03:01) & P61769 (B2M)).
Catalog No. 50-233-1742